1-Bromo-2,5-dichloro-3-fluorobenzene 98%

Supplier: Ambeed
Warning

A272913-1G A272913-25G A272913-100G A272913-10G A272913-500G
103813-116EA 5 USD
103813-116 103853-030 77092-818 77092-820 77153-650 77153-652 77153-654
1-Bromo-2,5-dichloro-3-fluorobenzene 98%
1-Bromo-2,5-dichloro-3-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂F
MW: 243.89 g/mol
Boiling Pt: 492.6 °C (760 mm Hg)
Flash Pt: 94.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00142585
CAS Number: 202865-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR