1-Bromo-2,4,5-trifluorobenzene 98%

Supplier: Ambeed
Danger

A734309-25G A734309-5G A734309-10G
103840-802EA 6 USD
103840-802 76845-030 76845-032 77105-126
1-Bromo-2,4,5-trifluorobenzene 98%
1-Bromo-2,4,5-trifluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₃
Boiling Pt: 144 °C (760 mm Hg)
Melting Pt: 19 °C
Flash Pt: 56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000306
CAS Number: 327-52-6
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR