1-Bromo-2,4,5-trichlorobenzene 97%

Supplier: Ambeed
Warning

A568463-25G A568463-5G A568463-100G A568463-10G A568463-1G
103828-794EA 9 USD
103828-794 103839-936 76844-062 76844-064 77103-670 77103-672 77307-840
1-Bromo-2,4,5-trichlorobenzene 97%
1-Bromo-2,4,5-trichlorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₃
MW: 260.34 g/mol
Boiling Pt: 268.9 °C (760 mm Hg)
Melting Pt: 139 °C
Flash Pt: 128.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00155012
CAS Number: 29682-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR