3-Bromoveratrole 95%

Supplier: Ambeed
Warning

A306055-5G A306055-100MG A306055-10G A306055-250MG A306055-1G
103814-452EA 24 USD
103814-452 103835-620 76838-324 76977-800 76977-802 76977-804 77305-930
3-Bromoveratrole 95%
3-Bromoveratrole

Precaución: For research use only

Formula: C₈H₉BrO₂
Boiling Pt: 228.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00185638
CAS Number: 5424-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR