1-Bromo-2,3-difluorobenzene 97%

Supplier: Ambeed
Warning

A161696-100G A161696-10G A161696-500G A161696-1G A161696-25G A161696-5G
103805-522EA 7 USD
103805-522 103843-764 77096-186 77096-188 77096-190 77319-756 77319-758 77319-760
1-Bromo-2,3-difluorobenzene 97%
1-Bromo-2,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrF₂
Boiling Pt: 158.3 °C (760 mm Hg)
Flash Pt: 61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00061136
CAS Number: 38573-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR