1-Bromo-2,3-dichloro-5-fluorobenzene 97%

Supplier: Ambeed
Warning

A417608-1G A417608-100MG A417608-250MG A417608-5G
103827-336EA 33 USD
103827-336 103838-382 76841-972 77115-898 77115-900 77307-188
1-Bromo-2,3-dichloro-5-fluorobenzene 97%
1-Bromo-2,3-dichloro-5-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂F
Storage Temperature: Ambient
MDL Number: MFCD09878107
CAS Number: 1000577-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR