1-Bromo-2,3,5-trichlorobenzene 96%

Supplier: Ambeed
Warning

A125862-100G A125862-10G A125862-25G A125862-5G
77234-312EA 192.61 USD
77234-312 77234-314 77312-886 77312-888
1-Bromo-2,3,5-trichlorobenzene 96%
1-Bromo-2,3,5-trichlorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₃
MW: 260.34 g/mol
Boiling Pt: 270.8 °C (760 mm Hg)
Melting Pt: 58…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00070734
CAS Number: 81067-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR