Bromopentafluorobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2,3,4,5,6-pentafluorobenzene

A144736-100G A144736-10G A144736-500G A144736-5G A144736-1000G A144736-25G
103808-652EA 226 USD
103808-652 76689-124 76689-126 76689-128 76689-130 77316-768 77316-770
Bromopentafluorobenzene 98%
Bromopentafluorobenzene

Precaución: for Research Use Only

Formula: C₆BrF₅
MW: 246.96 g/mol
Boiling Pt: 136.4 °C (760 mm Hg)
Melting Pt: …31 °C
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00000287
CAS Number: 344-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR