1-Boc-5-aminoindole 95%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-1H-indole, N1-BOC protected, 1-Boc-5-Aminoindole

A167142-100MG A167142-250MG A167142-25G A167142-5G A167142-1G
103806-136EA 16 USD
103806-136 103844-398 76720-362 76720-364 77081-392 77081-394 77300-874
1-Boc-5-aminoindole 95%
1-Boc-5-aminoindole

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₂
MW: 232.28
Boiling Pt: 384.1 °C (760 mm Hg)
Flash Pt: 186.128 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114746
CAS Number: 166104-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR