1-Boc-4-methylpiperidine-4-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-4-Methylpiperidine-4-carboxylic acid, 1-[(tert-butoxy)carbonyl]-4-methylpiperidine-4-carboxylic acid

A440349-1G A440349-5G A440349-10G A440349-250MG A440349-25G
103827-660EA 16 USD
103827-660 103838-726 76842-486 76842-488 77102-428 77102-430 77307-308
1-Boc-4-methylpiperidine-4-carboxylic acid 97%
1-Boc-4-methylpiperidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MW: 243.3000 g/mol
Boiling Pt: 354.3 °C (760 mm Hg)
Flash Pt: 168.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179132
CAS Number: 189321-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR