1-Boc4-hydroxy-3-methylpiperidine 95%

Supplier: Ambeed
Warning

A111344-250MG A111344-5G
103797-824EA 79 USD
103797-824 103823-640 76970-818 77310-448
1-Boc4-hydroxy-3-methylpiperidine 95%
1-Boc4-hydroxy-3-methylpiperidine

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
MW: 215.29 g/mol
Boiling Pt: 301.4 °C (760 mm Hg)
Flash Pt: 136.1 °C
Storage Temperature: Ambient
MDL Number: MFCD09701318
CAS Number: 181269-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR