tert-Butyl 4-fluoro-4-(hydroxymethyl)piperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-4-Fluoro-4-(hydroxymethyl)piperidine

A141610-250MG A141610-1G A141610-5G A141610-25G
103803-024EA 11 USD
103803-024 77027-582 77316-192 77316-194 77871-683
tert-Butyl 4-fluoro-4-(hydroxymethyl)piperidine-1-carboxylate 95%
tert-Butyl 4-fluoro-4-(hydroxymethyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₀FNO₃
Boiling Pt: 311 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08062513
CAS Number: 614730-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR