1-Boc-4-cyanopiperidine 98%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl 4-cyanopiperidine-1-carboxylate, N-BOC-Piperidine-4-carbonitrile

A117423-100G A117423-10G A117423-25G A117423-500G A117423-1G A117423-5G
103798-884EA 6 USD
103798-884 103824-768 76797-150 76797-152 76797-154 76797-156 77311-550 77311-552
1-Boc-4-cyanopiperidine 98%
1-Boc-4-cyanopiperidine

Precaución: for Research Use Only

Formula: C₁₁H₁₈N₂O₂
Melting Pt: 60…63 °C
Flash Pt: 150.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01861223
CAS Number: 91419-52-2
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR