1-Boc-4-chlorosulphonylpiperidine 98%

Supplier: Ambeed
Danger

Synonyms: 1-Boc-4-Chlorosulphonylpiperidine, 1-BOC-4-PIPERIDINEsulphONYL CHLORIDE, tert-Butyl 4-(chlorosulphonyl)piperidine-1-carboxylate, 4-CHLOROsulphONYL-PIPERIDINE-1-CARBOXYLIC acid TERT-BUTYL ESTER

A253329-100MG A253329-250MG A253329-1G A253329-10G
103812-090EA 25 USD
103812-090 103851-938 76786-846 76786-848 77091-370 77568-456
1-Boc-4-chlorosulphonylpiperidine 98%
1-Boc-4-chlorosulfonylpiperidine

Precaución: For research use only.

Formula: C₁₀H₁₈ClNO₄S
Boiling Pt: 369.5 °C (760 mm Hg)
Melting Pt: 83…87 °C
Flash Pt: 177.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01861750
CAS Number: 782501-25-1
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR