4-Amino-1-boc-piperidine-4-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-1-(tert-butoxycarbonyl)piperidine-4-carboxylic acid

A319862-10G A319862-250MG A319862-1G A319862-25G A319862-5G A319862-100G
103825-222EA 14 USD
103825-222 103836-154 76802-890 76802-892 76839-012 76839-014 76839-016 77668-422
4-Amino-1-boc-piperidine-4-carboxylic acid 97%
4-Amino-1-Boc-piperidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₄
MW: 244.29 g/mol
Boiling Pt: 380.1 °C (760 mm Hg)
Melting Pt: 290 °C
Flash Pt: 183.7 °C
Storage Temperature: Ambient
MDL Number: MFCD01318728
CAS Number: 183673-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR