1-Boc-4-(3-formylphenyl)piperazine 95%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-4-(3-Formylphenyl)piperazine

A175868-250MG A175868-1G
103807-092EA 77 USD
103807-092 103845-404 77029-250 77301-404
1-Boc-4-(3-formylphenyl)piperazine 95%
1-Boc-4-(3-formylphenyl)piperazine

Precaución: for Research Use Only

Formula: C₁₆H₂₂N₂O₃
MW: 290.3600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06657284
CAS Number: 1257849-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR