1-Boc-4-(3-aminopyridin-2-yl)piperazine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Boc-4-(3-Aminopyridin-2-yl)piperazine
A238376-1G
A238376-5G
A238376-10G
A238376-250MG
103811-226EA
115
USD
103811-226
103851-006
77090-120
77090-122
77275-952
77275-954
1-Boc-4-(3-aminopyridin-2-yl)piperazine 97%
1-Boc-4-(3-aminopyridin-2-yl)piperazine
Precaución: for Research Use Only
Formula:
C₁₄H₂₂N₄O₂ MW: 278.3501 g/mol Boiling Pt: 448.8 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD03407996 CAS Number: 111669-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...