tert-Butyl-4-(3-ethoxy-3-oxopropanoyl)tetrahydro-1(2H)-pyridinecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-4-(2-Ethoxycarbonyl-acetyl)piperidine

A292184-1G A292184-25G A292184-5G A292184-100G A292184-10G A292184-250MG
103813-930EA 16 USD
103813-930 103835-070 76837-664 76837-666 76837-668 77289-800 77289-802 77289-804
tert-Butyl-4-(3-ethoxy-3-oxopropanoyl)tetrahydro-1(2H)-pyridinecarboxylate 97%
tert-Butyl-4-(3-ethoxy-3-oxopropanoyl)tetrahydro-1(2H)-pyridinecarboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₅NO₅
MW: 299.37 g/mol
Boiling Pt: 386.1 °C (760 mm Hg)
Flash Pt: 187.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115569
CAS Number: 479630-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR