1-Boc-3-pyrrolidineacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-3-Pyrrolidineacetic acid

A102223-10G A102223-250MG A102223-25G A102223-1G A102223-5G
103796-338EA 14 USD
103796-338 103822-032 77093-562 77093-564 77093-566 77308-908 77308-910
1-Boc-3-pyrrolidineacetic acid 97%
1-Boc-3-pyrrolidineacetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.2700 g/mol
Boiling Pt: 357.4 °C (760 mm Hg)
Flash Pt: 169.945 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179024
CAS Number: 175526-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR