1-Boc-3-piperidone 98%

Supplier: Ambeed
Warning

Synonyms: N-BOC-3-piperidone, 1-Boc-3-Piperidinone

A239087-100G A239087-10G A239087-1G A239087-500G A239087-25G A239087-5G
103851-060EA 6 USD
103851-060 76771-358 76771-360 76771-362 76771-364 77090-196 77090-198
1-Boc-3-piperidone 98%
1-Boc-3-piperidone

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₃
MW: 199.25 g/mol
Melting Pt: 35…40 °C
Flash Pt: 110 °C
Storage Temperature: Freezer
MDL Number: MFCD01631193
CAS Number: 98977-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR