1-Boc-3-oxopiperazine 97%

Supplier: Ambeed
Warning

Synonyms: tert-butyl 3-oxopiperazine-1-carboxylate, tert-butyl 3-oxo-1-piperazinecarboxylate, 1-Boc-3-Oxopiperazine, 4-N-BOC-2-oxo-piperazine, 4-Boc-2-Oxopiperazine

A320272-25G A320272-5G A320272-10G A320272-500G A320272-100G
103836-162EA 5 USD
103836-162 76839-028 76839-030 77263-394 77263-396 77593-896
1-Boc-3-oxopiperazine 97%
1-Boc-3-oxopiperazine

Precaución: for Research Use Only

Formula: C₉H₁₆N₂O₃
Melting Pt: 156…160 °C
Flash Pt: 171 °C
Storage Temperature: Ambient
MDL Number: MFCD02181069
CAS Number: 76003-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR