1-Boc-3-Oxo-1,4-diazepane 95%

Supplier: Ambeed
Warning

A162245-100MG A162245-250MG
103805-594EA 100 USD
103805-594 103843-844 77028-656 77028-658
1-Boc-3-Oxo-1,4-diazepane 95%
1-Boc-3-Oxo-1,4-diazepane

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₃
MW: 214.26 g/mol
Boiling Pt: 373.1 °C (760 mm Hg)
Flash Pt: 179.4 °C
Storage Temperature: Ambient
MDL Number: MFCD11975719
CAS Number: 179686-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR