(±)-1-Boc-3-(methylamino)pyrrolidine 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-(methylamino)pyrrolidine-1-carboxylate

A137902-100G A137902-250MG A137902-25G A137902-1G A137902-5G
103802-526EA 10 USD
103802-526 76972-434 76972-436 76972-438 77315-522 77315-524
(±)-1-Boc-3-(methylamino)pyrrolidine 97%
(±)-1-Boc-3-(methylamino)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MW: 200.2780 g/mol
Boiling Pt: 269.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06656606
CAS Number: 454712-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR