tert-Butyl-3-[(methylsulfonyl)oxy]-1-azetanecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-3-Methanesulphonyloxyazetidine

A454215-1G A454215-25G A454215-5G A454215-100G A454215-10G A454215-250MG
103827-822EA 9 USD
103827-822 103838-892 76842-730 76842-732 76842-734 77196-508 77196-510 77196-512
tert-Butyl-3-[(methylsulfonyl)oxy]-1-azetanecarboxylate 98%
tert-Butyl-3-[(methylsulfonyl)oxy]-1-azetanecarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₇NO₅S
MW: 251.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05664832
CAS Number: 141699-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR