1-Boc-3-hydroxymethylindole 97%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-3-Hydroxymethylindole, 3-(Hydroxymethyl)-1H-indole, N-BOC protected

A117660-250MG A117660-25G
103798-922EA 8 USD
103798-922 103824-810 77148-980 77148-982
1-Boc-3-hydroxymethylindole 97%
1-Boc-3-hydroxymethylindole

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₃
MW: 247.2900 g/mol
Boiling Pt: 396 °C (760 mm Hg)
Flash Pt: 193.3 °C
Storage Temperature: Ambient
MDL Number: MFCD05864717
CAS Number: 96551-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR