1-Boc-3-ethynylpyrrolidine 95%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-3-Ethynylpyrrolidine

A307881-1G A307881-5G A307881-10G A307881-250MG
103814-528EA 81 USD
103814-528 103835-704 76838-448 76838-450 76875-732 76875-734
1-Boc-3-ethynylpyrrolidine 95%
1-Boc-3-ethynylpyrrolidine

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₂
MW: 195.26 g/mol
Flash Pt: 104.491 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10698160
CAS Number: 287193-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR