1-Boc-3-carbamoylpiperidine 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-Carbamoyl-1-piperidinecarboxylate, 1-Boc-3-piperidinecarboxamide, 3-Carbamoyl-1-piperidinecarboxylic acid tert-Butyl ester, 1-Boc-3-Carbamoylpiperidine, 1-(tert-Butoxycarbonyl)-3-piperidinecarboxamide

A166283-10G A166283-25G A166283-1G A166283-5G
103806-042EA 7 USD
103806-042 103844-302 76872-194 76872-196 77320-546 77320-548
1-Boc-3-carbamoylpiperidine 95%
1-Boc-3-carbamoylpiperidine

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₃
MW: 228.2881 g/mol
Boiling Pt: 384.4 °C (760 mm Hg)
Melting Pt: 170…176 °C
Storage Temperature: Ambient
MDL Number: MFCD06658365
CAS Number: 91419-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR