(±)-1-Boc-3-aminopyrrolidine 97%
Supplier: Ambeed
|
Danger
|
A158386-10G
A158386-1G
A158386-25G
A158386-5G
103805-126EA
12
USD
103805-126
103843-336
77259-498
77300-326
77319-194
77319-196
(±)-1-Boc-3-aminopyrrolidine 97%
(±)-1-Boc-3-aminopyrrolidine
Precaución: for Research Use Only
Formula:
C₉H₁₈N₂O₂ Boiling Pt: 257.4 °C (760 mm Hg) Flash Pt: 91 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01861220 CAS Number: 186550-13-0 UN: 2922 ADR: 8,6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...