1-Boc-3,3-dimethylpiperazine 98%

Supplier: Ambeed
Warning

A144839-100MG A144839-10G A144839-1G A144839-5G
103803-458EA 16 USD
103803-458 103841-536 76984-398 76984-400 77299-416 77299-418
1-Boc-3,3-dimethylpiperazine 98%
1-Boc-3,3-dimethylpiperazine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD07371651
CAS Number: 259808-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR