1-Boc-2,6-dimethylpiperazine 95%

Supplier: Ambeed
Warning

A166852-100MG A166852-250MG A166852-5G A166852-1G
103806-104EA 35 USD
103806-104 103844-366 76985-246 76985-248 77300-854 77320-644
1-Boc-2,6-dimethylpiperazine 95%
1-Boc-2,6-dimethylpiperazine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07371653
CAS Number: 688363-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR