tert-Butyl 1,8-diazaspiro[4.5]decane-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-1,8-diaza-spiro[4.5]decane

A259092-100MG A259092-250MG A259092-1G A259092-5G
103812-422EA 208 USD
103812-422 103852-298 76988-146 76988-148 77091-834 77304-762
tert-Butyl 1,8-diazaspiro[4.5]decane-1-carboxylate 95%
tert-Butyl 1,8-diazaspiro[4.5]decane-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₂
MW: 240.342 g/mol
Boiling Pt: 337 °C (760 mm Hg)
Flash Pt: 158 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06739037
CAS Number: 885279-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR