1-Benzylpyrrolidine-2,5-dione 97%

Supplier: Ambeed
Warning

A355672-1G A355672-5G A355672-10G A355672-250MG
103826-148EA 11 USD
103826-148 103837-154 76840-280 76840-282 77033-148 77033-150
1-Benzylpyrrolidine-2,5-dione 97%
1-Benzylpyrrolidine-2,5-dione

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MW: 189.211 g/mol
Boiling Pt: 394.5 °C (760 mm Hg)
Melting Pt: 100…103 °C
Flash Pt: 194.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00516834
CAS Number: 2142-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR