1-Benzyl-pyrrolidine 95%

Supplier: Ambeed
Warning

Synonyms: 1-Benzylpyrrolidine

A383339-10G A383339-250MG A383339-1G A383339-5G
103826-734EA 14 USD
103826-734 103837-770 76724-900 76724-902 76841-094 76841-096
1-Benzyl-pyrrolidine 95%
1-Benzyl-pyrrolidine

Precaución: for Research Use Only

Formula: C₁₁H₁₅N
MW: 161.244 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00127807
CAS Number: 29897-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR