1-Benzylpiperidine-4-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A253334-10G
A253334-250MG
A253334-1G
A253334-25G
A253334-5G
103812-092EA
9
USD
103812-092
103851-940
77014-038
77014-040
77091-372
77091-374
77091-376
1-Benzylpiperidine-4-carboxylic acid 97%
1-Benzylpiperidine-4-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₃H₁₇NO₂ Boiling Pt: 358.6 °C (760 mm Hg) Flash Pt: 170.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03371463 CAS Number: 10315-07-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...