1-Benzylpiperidine-4-carbonitrile 97%

Supplier: Ambeed
Warning

A126015-10G A126015-1G A126015-5G
103800-752EA 30 USD
103800-752 76782-014 77298-104 77298-106
1-Benzylpiperidine-4-carbonitrile 97%
1-Benzylpiperidine-4-carbonitrile

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂
MW: 200.2795 g/mol
Boiling Pt: 330.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01540574
CAS Number: 62718-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR