1-Benzylpiperidine 98%

Supplier: Ambeed
Warning

A624079-1G A624079-25G A624079-5G A624079-100G
103840-236EA 22 USD
103840-236 76844-404 76844-406 76844-408 77198-238
1-Benzylpiperidine 98%
1-Benzylpiperidine

Precaución: for Research Use Only

Formula: C₁₂H₁₇N
MW: 175.28 g/mol
Boiling Pt: 120…123 °C at 9 mmHg
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00224901
CAS Number: 2905-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR