1-Benzyl-5-nitro-1H-indazole 98%

Supplier: Ambeed
Warning

A101064-250MG A101064-1G A101064-5G
103795-786EA 47 USD
103795-786 103821-476 76780-446 77296-970 77296-972
1-Benzyl-5-nitro-1H-indazole 98%
1-Benzyl-5-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₁₄H₁₁N₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD08275706
CAS Number: 23856-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR