1-Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine 95%
Supplier: Ambeed
|
Warning
|
A183372-100MG
A183372-250MG
A183372-1G
103815-636EA
13
USD
103815-636
103846-198
77029-528
77029-530
77083-802
1-Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine 95%
1-Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine
Precaución: for Research Use Only
Formula:
C₁₈H₂₆BNO₂ MW: 299.216 g/mol Boiling Pt: 357.7 °C (760 mm Hg) Storage Temperature: Freezer |
MDL Number:
MFCD11501654 CAS Number: 1048976-83-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...