1-Benzyl-3-pyrrolidinone 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(Phenylmethyl)-3-pyrrolidinone, 1-Benzylpyrrolidin-3-one
A694341-1G
A694341-5G
A694341-100G
103840-602EA
9
USD
103840-602
77280-784
77308-138
77749-428
1-Benzyl-3-pyrrolidinone 97%
1-Benzyl-3-pyrrolidinone
Precaución: for Research Use Only
Formula:
C₁₁H₁₃NO Boiling Pt: 77 °C at 0.01 mmHg Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005342 CAS Number: 775-16-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...