1-Benzyl-3,3-difluoropiperidin-4-one 97%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl-3,3-difluoro-4-oxopiperidine

A109947-100MG A109947-250MG A109947-25G A109947-1G A109947-5G
103797-592EA 63 USD
103797-592 103823-400 76982-674 76982-676 76982-678 77310-206 77310-208
1-Benzyl-3,3-difluoropiperidin-4-one 97%
1-Benzyl-3,3-difluoropiperidin-4-one

Precaución: for Research Use Only

Formula: C₁₂H₁₃F₂NO
MW: 225.235 g/mol
Boiling Pt: 291.0±40.0 °C (760 mm Hg)
Flash Pt: 129.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD15526740
CAS Number: 1039741-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR