1-Benzyl-1H-pyrazol-4-amine hydrochloride 97%

Supplier: Ambeed
Warning

A101884-1G A101884-250MG
103796-196EA 12 USD
103796-196 103821-890 77308-778 77745-777
1-Benzyl-1H-pyrazol-4-amine hydrochloride 97%
1-Benzyl-1H-pyrazol-4-amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₁N₃·ClH
MW: 209.675 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18425906
CAS Number: 1264097-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR