1-Benzyl-1H-pyrazol-4-amine 97%

Supplier: Ambeed
Warning

A141560-250MG A141560-5G
103803-014EA 85 USD
103803-014 76996-886 77316-180
1-Benzyl-1H-pyrazol-4-amine 97%
1-Benzyl-1H-pyrazol-4-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₁N₃
Boiling Pt: 369.4 °C (760 mm Hg)
Flash Pt: 177.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01693690
CAS Number: 28466-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR