1-Benzylimidazole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Benzyl-1H-imidazole
A135013-10G
A135013-1G
A135013-100G
A135013-500G
103802-082EA
5
USD
103802-082
77137-056
77314-862
77685-163
77685-164
1-Benzylimidazole 98%
1-Benzylimidazole
Precaución: for Research Use Only
Formula:
C₁₀H₁₀N₂ MW: 158.2 g/mol Boiling Pt: 310 Melting Pt: 72 Flash Pt: 141.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005296 CAS Number: 4238-71-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...