1-Benzyl 3-methyl 4-oxopiperidine-1,3-dicarboxylate 98%

Supplier: Ambeed
Warning

A382644-1G A382644-5G
103826-720EA 68 USD
103826-720 103837-756 76841-078 77306-938
1-Benzyl 3-methyl 4-oxopiperidine-1,3-dicarboxylate 98%
1-Benzyl 3-methyl 4-oxopiperidine-1,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₇NO₅
MW: 291.29 g/mol
Boiling Pt: 435.2 °C (760 mm Hg)
Flash Pt: 217 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09910357
CAS Number: 159299-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR