1-Benzoylpiperidine-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A104256-250MG A104256-25G A104256-1G A104256-5G
103796-674EA 9 USD
103796-674 103822-384 76717-700 76717-702 77309-250 77309-252
1-Benzoylpiperidine-4-carboxylic acid 98%
1-Benzoylpiperidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₃
Boiling Pt: 438.8 °C (760 mm Hg)
Melting Pt: 144…146 °C
Flash Pt: 219.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00040747
CAS Number: 5274-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR