Benzo[b]thiophene-3-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Benzothiophene-3-carboxylic acid
A198185-10G
A198185-1G
A198185-5G
103816-996EA
11
USD
103816-996
103847-622
76691-116
76691-118
77085-636
77085-638
Benzo[b]thiophene-3-carboxylic acid 97%
Benzo[b]thiophene-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₉H₆O₂S Boiling Pt: 376.2 °C (760 mm Hg) Melting Pt: 174…176 °C Flash Pt: 181.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD01846406 CAS Number: 5381-25-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...