Aminoacetone hydrochloride 95%

Supplier: Ambeed
Warning

A135066-250MG A135066-25G A135066-1G A135066-5G A135066-100G
103802-086EA 14 USD
103802-086 76688-774 76688-776 77314-868 77314-870 77624-134
Aminoacetone hydrochloride 95%
Aminoacetone hydrochloride

Precaución: for Research Use Only

Formula: C₃H₈ClNO
Melting Pt: 73…77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00056125
CAS Number: 7737-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR