1H-Indol-1-amine 97%

Supplier: Ambeed
Warning

Synonyms: 1-Aminoindole

A434542-100MG A434542-1G A434542-5G
103827-576EA 36 USD
103827-576 103838-642 77116-100 77307-278 77307-280
1H-Indol-1-amine 97%
1H-Indol-1-amine

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.163 g/mol
Boiling Pt: 296.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00151983
CAS Number: 53406-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR