1-Amino-9,10-anthraquinone 95%

Supplier: Ambeed
Warning

Synonyms: 1-Aminoanthraquinone, 1-Aminoanthracene-9,10-dione

A202774-25G A202774-100G A202774-500G
103809-376EA 89 USD
103809-376 77086-174 77192-378 77192-380
1-Amino-9,10-anthraquinone 95%
1-Amino-9,10-anthraquinone

Precaución: for Research Use Only

Formula: C₁₄H₉NO₂
MW: 223.23 g/mol
Boiling Pt: 464.9 °C (760 mm Hg)
Melting Pt: 253…255 °C
Flash Pt: 235 °C
Storage Temperature: Ambient
MDL Number: MFCD00001213
CAS Number: 82-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR