1-Amino-2-methylpropan-2-ol 98%
Supplier: Ambeed
|
Danger
|
A260796-25G
A260796-5G
A260796-100G
A260796-10G
A260796-1G
103812-506EA
7
USD
103812-506
103852-392
77091-982
77091-984
77193-812
77193-814
77304-804
1-Amino-2-methylpropan-2-ol 98%
1-Amino-2-methylpropan-2-ol
Precaución: for Research Use Only
Formula:
C₄H₁₁NO Boiling Pt: 167.2 °C (760 mm Hg) Flash Pt: 73.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01697283 CAS Number: 2854-16-2 UN: 2924 ADR: 3,8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...